Assay Details
| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172280.2 | NP_758484.2 | 18 - 19 | 3257 | 95 | |
| GenBank mRNA | BC020109.1 | - | 3 - 4 | 758 | 95 | |
| BC089304.1 | - | 18 - 19 | 3257 | 6854147 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

2210018M11Rik
Genomic Map