Assay Details
| Gene Symbol: Slco2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252530.1 | NP_001239459.1 | 11 - 12 | 2075 | 67 | |
| NM_001252531.1 | NP_001239460.1 | 11 - 12 | 2018 | 67 | ||
| NM_175316.3 | NP_780525.1 | 11 - 12 | 1821 | 67 | ||
| GenBank mRNA | AK033539.1 | - | 3 - 4 | 447 | 67 | |
| AK048120.1 | - | 11 - 12 | 1822 | 67 | ||
| AK087682.1 | - | 11 - 12 | 2073 | 67 | ||
| AK132394.1 | - | 11 - 12 | 2076 | 67 | ||
| AK163542.1 | - | 11 - 12 | 2071 | 67 | ||
| AK173049.1 | - | 9 - 10 | 1607 | 67 | ||
| BC019209.1 | - | 11 - 12 | 1804 | 67 | ||
| BC037089.1 | - | 11 - 12 | 1793 | 67 | ||
| BC049251.1 | - | 11 - 12 | 1784 | 67 | ||
| BC096485.1 | - | 11 - 12 | 1830 | 5101188 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slco2b1
Genomic Map