Assay Details
| Gene Symbol: Slco2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252530.1 | NP_001239459.1 | 4 - 5 | 770 | 71 | |
| NM_001252531.1 | NP_001239460.1 | 4 - 5 | 713 | 71 | ||
| NM_175316.3 | NP_780525.1 | 4 - 5 | 516 | 71 | ||
| GenBank mRNA | AK041144.1 | - | 4 - 5 | 712 | 71 | |
| AK048120.1 | - | 4 - 5 | 517 | 71 | ||
| AK087682.1 | - | 4 - 5 | 764 | 71 | ||
| AK132394.1 | - | 4 - 5 | 771 | 71 | ||
| AK163542.1 | - | 4 - 5 | 766 | 71 | ||
| AK173049.1 | - | 2 - 3 | 302 | 71 | ||
| BC019209.1 | - | 4 - 5 | 499 | 71 | ||
| BC037089.1 | - | 4 - 5 | 488 | 71 | ||
| BC049251.1 | - | 4 - 5 | 479 | 71 | ||
| BC096485.1 | - | 4 - 5 | 525 | 5101188 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slco2b1
Genomic Map