Assay Details
| Gene Symbol: Arhgap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287530.1 | NP_001274459.1 | 10 - 11 | 1671 | 84 | |
| NM_009707.4 | NP_033837.2 | 10 - 11 | 3039 | 84 | ||
| NM_178754.3 | NP_848869.1 | 10 - 11 | 1582 | 84 | ||
| GenBank mRNA | AF012273.2 | - | 2161 | 84 | ||
| AK048162.1 | - | 10 - 11 | 1536 | 84 | ||
| AK048507.1 | - | 10 - 11 | 1537 | 84 | ||
| AK077996.1 | - | 10 - 11 | 1544 | 84 | ||
| AK166015.1 | - | 10 - 11 | 1502 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap6
Genomic Map