Assay Details
| Gene Symbol: Cttnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080285.1 | NP_525024.1 | 11 - 12 | 3324 | 98 | |
| GenBank mRNA | AK032356.1 | - | 9 - 10 | 1705 | 98 | |
| AK081137.1 | - | 7 - 8 | 1787 | 98 | ||
| AK173254.1 | - | 11 - 12 | 3230 | 98 | ||
| BC068156.1 | - | 11 - 12 | 3085 | 98 | ||
| BC141407.1 | - | 11 - 12 | 3320 | 9056020 | 98 | |
More Information
Set Membership: |
|

Cttnbp2
Genomic Map