Assay Details
| Gene Symbol: Smpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045569.1 | 3 - 4 | 438 | 86 | ||
| GenBank mRNA | W18646.1 | - | 3 - 4 | 151 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smpx
Genomic Map