Assay Details
| Gene Symbol: Mtdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026002.4 | NP_080278.3 | 4 - 5 | 1059 | 99 | |
| GenBank mRNA | AF501309.1 | - | 4 - 5 | 997 | 99 | |
| AK029915.1 | - | 4 - 5 | 1060 | 99 | ||
| AK151269.1 | - | 4 - 5 | 1043 | 99 | ||
| AY553638.1 | - | 4 - 5 | 742 | 99 | ||
| BC023115.1 | - | 4 - 5 | 958 | 99 | ||
| BC036994.1 | - | 4 - 5 | 778 | 99 | ||
| BC138858.1 | - | 4 - 5 | 773 | 8861880 | 99 | |
| BC138859.1 | - | 4 - 5 | 773 | 8861881 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mtdh
Genomic Map