Assay Details
| Gene Symbol: Scnn1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011326.2 | NP_035456.1 | 3 - 4 | 701 | 67 | |
| GenBank mRNA | AF112187.1 | - | 3 - 4 | 701 | 67 | |
| AK143995.1 | - | 2 - 3 | 394 | 67 | ||
| BC021338.1 | - | 3 - 4 | 740 | 5042876 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scnn1g
Genomic Map