Assay Details
| Gene Symbol: Irf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252600.1 | NP_001239529.1 | 6 - 7 | 1062 | 112 | |
| GenBank mRNA | AK152403.1 | - | 6 - 7 | 1062 | 112 | |
| AK159883.1 | - | 6 - 7 | 1062 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf7
Genomic Map