Assay Details
| Gene Symbol: Irf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252600.1 | NP_001239529.1 | 7 - 8 | 1445 | 144 | |
| NM_001252601.1 | NP_001239530.1 | 8 - 9 | 1442 | 144 | ||
| NM_016850.3 | NP_058546.1 | 8 - 9 | 1538 | 144 | ||
| GenBank mRNA | AK002830.1 | - | 8 - 9 | 1440 | 144 | |
| AK004349.1 | - | 8 - 9 | 1537 | 144 | ||
| AK079685.1 | - | 8 - 9 | 1538 | 144 | ||
| AK150403.1 | - | 8 - 9 | 1538 | 144 | ||
| AK150608.1 | - | 8 - 9 | 1538 | 144 | ||
| AK151470.1 | - | 8 - 9 | 1539 | 144 | ||
| AK152401.1 | - | 8 - 9 | 1538 | 144 | ||
| AK152403.1 | - | 7 - 8 | 1445 | 144 | ||
| AK153054.1 | - | 8 - 9 | 1538 | 144 | ||
| AK158378.1 | - | 8 - 9 | 1442 | 144 | ||
| AK158403.1 | - | 8 - 9 | 1442 | 144 | ||
| AK159142.1 | - | 8 - 9 | 1537 | 144 | ||
| AK159883.1 | - | 7 - 8 | 1445 | 144 | ||
| BC138799.1 | - | 7 - 8 | 1478 | 8861821 | 144 | |
| BC145422.1 | - | 6 - 7 | 1323 | 144 | ||
| U73037.1 | - | 7 - 8 | 1515 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf7
Genomic Map