Assay Details
| Gene Symbol: Sat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291865.1 | NP_001278794.1 | 4 - 5 | 589 | 115 | |
| GenBank mRNA | BC108334.1 | - | 3 - 4 | 790 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001291865.1 transcripts.
This assay is in the 5'UTR region of the NM_001291865.1 transcripts.
Set Membership: |
|

Sat1
Genomic Map