Assay Details
| Gene Symbol: Prkag2os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040684.1 | 2 - 3 | 389 | 63 | ||
| GenBank mRNA | AK016926.1 | - | 2 - 3 | 388 | 63 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY716606.1 | - | 385 | Chr. - - [-] on Build | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Ras homolog enriched in brain | Rheb | Rheb1 | -60.8kb | Anti-sense | Chr. 5 - 24802823 - 24842361 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1500035N22 gene | 1500035N22Rik | 80.1kb | Sense | Chr. 5 - 24984548 - 24998560 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Prkag2os1
Genomic Map