Assay Details
| Gene Symbol: Gm38793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK085647.1 | - | 244 | 160 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
thromboxane A synthase 1, platelet |
Tbxas1 |
CYP5, CYP5A1, THAS, TS, TXAS, TXS |
Anti-sense | Chr. 6 - 38918467 - 39084590 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 1700025N23 gene | 1700025N23Rik | 88.3kb | Sense | Chr. 6 - 39063879 - 39067586 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm38793
Genomic Map