Assay Details
| Gene Symbol: 4921504A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102341.1 | 3 - 4 | 599 | 115 | ||
| GenBank mRNA | AK046351.1 | - | 3 - 4 | 599 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31752 |
Gm31752 | -707.1kb | Sense | Chr. 5 - 18481690 - 18495260 on NCBI Build GRCm38 | |
| Downstream |
membrane associated guanylate kinase, WW and PDZ domain containing 2 |
Magi2 |
AIP-1, Acvri1, Acvrinp1, Acvrip1, Magi-2, S-SCAM, mKIAA0705 |
16.1kb | Anti-sense | Chr. 5 - 19223283 - 20704792 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4921504A21Rik
Genomic Map