Assay Details
| Gene Symbol: Gm31752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076902.1 | - | 6 - 7 | 585 | 152 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
guanine nucleotide binding protein (G protein), alpha inhibiting 1 |
Gnai1 |
AU046200, Gialpha1, Gnai-1 |
-121.3kb | Sense | Chr. 5 - 18265135 - 18360413 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4921504A21 gene | 4921504A21Rik | 718.7kb | Sense | Chr. 5 - 19202368 - 19226555 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31752
Genomic Map