Assay Details
| Gene Symbol: Grip1os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045359.1 | 2 - 3 | 181 | 64 | ||
| GenBank mRNA | AK016220.1 | - | 2 - 3 | 181 | 64 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY715827.1 | - | 181 | Chr. - - [-] on Build | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutamate receptor interacting protein 1 | Grip1 |
4931400F03Rik, GRIP, eb |
Anti-sense | Chr. 10 - 119453835 - 120087267 on NCBI Build GRCm38 | |
| Downstream |
glutamate receptor interacting protein 1, opposite strand 1 |
Grip1os1 |
4930404E01Rik, 4930483C13Rik |
63.4kb | Sense | Chr. 10 - 119823658 - 119873140 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Grip1os2
Genomic Map