Assay Details
| Gene Symbol: Fam19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252387.1 | NP_001239316.1 | 3 - 4 | 1725 | 75 | |
| NM_182807.3 | NP_877959.2 | 4 - 5 | 1937 | 75 | ||
| NR_045514.1 | 3 - 4 | 1725 | 75 | |||
| GenBank mRNA | AK032483.1 | - | 3 - 4 | 807 | 75 | |
| AK139403.1 | - | 3 - 4 | 1809 | 75 | ||
| AK140763.1 | - | 3 - 4 | 1688 | 75 | ||
| AK164201.1 | - | 3 - 4 | 817 | 75 | ||
| AY325121.1 | - | 4 - 5 | 624 | 75 | ||
| BC027082.1 | - | 4 - 5 | 1823 | 5360443 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam19a2
Genomic Map