Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Ras homolog enriched in brain | Rheb | Rheb1 | -32.2kb | Anti-sense | Chr. 5 - 24802823 - 24842361 on NCBI Build GRCm38 |
| Downstream |
protein kinase, AMP-activated, gamma 2 non-catalytic subunit, opposite strand 1 |
Prkag2os1 | 4933426D04Rik | 27.5kb | Sense | Chr. 5 - 24902569 - 24906849 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map