Assay Details
| Gene Symbol: 4930484H19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015621.1 | - | 2 - 3 | 1307 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 378d | Mir378d |
Gm27650, mmu-mir-378d |
-79.7kb | Sense | Chr. 10 - 126710282 - 126710391 on NCBI Build GRCm38 |
| Downstream | XRCC6 binding protein 1 | Xrcc6bp1 |
1110068E08Rik, 2410012H02Rik |
76.2kb | Sense | Chr. 10 - 126868427 - 126901711 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930484H19Rik
Genomic Map