Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
histone cluster 1, H2be |
Hist1h2be | Anti-sense | Chr. 13 - 23583670 - 23621124 on NCBI Build GRCm38 | ||
| Downstream |
histone cluster 1, H1e |
Hist1h1e |
H1-4, H1.4, H1e, H1f4, H1s-4, H1var2 |
2.4kb | Anti-sense | Chr. 13 - 23621777 - 23622558 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map