Assay Details
| Gene Symbol: Chpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133913.2 | NP_598674.2 | 2 - 3 | 2118 | 56 | |
| GenBank mRNA | AK089404.1 | - | 2 - 3 | 2128 | 56 | |
| AK138814.1 | - | 2124 | 56 | |||
| AK219661.1 | - | 1 - 2 | 50 | 56 | ||
| BC019714.1 | - | 1 - 2 | 179 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chpf2
Genomic Map