Assay Details
| Gene Symbol: Fastk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023229.2 | NP_075718.2 | 6 - 7 | 1273 | 102 | |
| GenBank mRNA | AF168682.1 | - | 5 - 6 | 1168 | 102 | |
| AK002552.1 | - | 6 - 7 | 1222 | 101 | ||
| AK003940.1 | - | 3 - 4 | 516 | 102 | ||
| AK038596.1 | - | 6 - 7 | 1225 | 102 | ||
| AK052667.1 | - | 6 - 7 | 1218 | 102 | ||
| AK139374.1 | - | 5 - 6 | 1993 | 102 | ||
| AK151859.1 | - | 6 - 7 | 1225 | 102 | ||
| AK152369.1 | - | 6 - 7 | 1274 | 102 | ||
| AK159751.1 | - | 6 - 7 | 1225 | 102 | ||
| AK164317.1 | - | 1231 | 102 | |||
| AK166508.1 | - | 1818 | 102 | |||
| AK167346.1 | - | 5 - 6 | 1985 | 102 | ||
| BC013547.1 | - | 6 - 7 | 1190 | 4218374 | 102 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fastk
Genomic Map