Assay Details
| Gene Symbol: Slc13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019481.2 | NP_062354.2 | 12 - 13 | 1367 | 153 | |
| GenBank mRNA | AK005153.1 | - | 6 - 7 | 701 | 153 | |
| AK128899.1 | - | 12 - 13 | 1367 | 153 | ||
| BC022672.1 | - | 8 - 9 | 746 | 153 | ||
| BC040789.1 | - | 12 - 13 | 1341 | 153 | ||
| BC049981.1 | - | 12 - 13 | 1349 | 5065026 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc13a1
Genomic Map