Assay Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028462.1 | NP_082738.1 | 1 - 2 | 991 | 77 | |
| GenBank mRNA | AK014242.1 | - | 1 - 2 | 869 | 77 | |
| BC049157.1 | - | 1 - 2 | 1090 | 77 | ||
| BC119565.1 | - | 1 - 2 | 991 | 40057718 | 77 | |
| BC119566.1 | - | 1 - 2 | 991 | 40057721 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neuron migration
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
nervous system development
axon guidance
negative regulation of cell proliferation
telencephalon development
olfactory bulb development
forebrain anterior/posterior pattern specification
cell differentiation
forebrain development
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
nucleic acid binding
DNA binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
neuron migration
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
nervous system development
axon guidance
negative regulation of cell proliferation
telencephalon development
olfactory bulb development
forebrain anterior/posterior pattern specification
cell differentiation
forebrain development
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
nucleic acid binding
DNA binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295387 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295388 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295392 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295397 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295381 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295386 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295389 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295390 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295383 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295400 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295385 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295384 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295395 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295396 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295391 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295401 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295399 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295393 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P347619 | NM_028462.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P347619 | AK014242.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P347619 | BC049157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P347619 | NM_028462.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P347619 | AK014242.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P347619 | BC049157.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 705527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102419 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135810 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P347619 | NM_028462.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P347619 | AK014242.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P347619 | BC049157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P347619 | NM_028462.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P347619 | AK014242.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P347619 | BC049157.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135810 | - | - |
| Whole Mouse Genome G4122A | A_51_P347619 | NM_028462.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P347619 | AK014242.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P347619 | BC049157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P347619 | NM_028462.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P347619 | AK014242.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P347619 | BC049157.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135810 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295397 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295385 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295384 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295396 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295434 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295435 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295433 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295399 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295389 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295388 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295400 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295401 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295383 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295386 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295395 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295393 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295392 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295391 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295387 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295390 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24295381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05372443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05267627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05549111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05372458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05207302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05372461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05068988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05549106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05207271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05207286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24295353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05267625 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10543365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543368 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429991_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10543362 | - | - |
| Mouse Genome 430 2.0 Array | 1429991_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429991_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73191_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73191_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600015100.mm.1 | NM_028462.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015100.mm.1 | NM_028462.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015100.mm.1 | TC0600001930.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600015102.mm.1 | NM_028462.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015102.mm.1 | NM_028462.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015102.mm.1 | TC0600001930.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600015103.mm.1 | NM_028462.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015103.mm.1 | NM_028462.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015103.mm.1 | TC0600001930.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600015101.mm.1 | NM_028462.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015101.mm.1 | NM_028462.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015101.mm.1 | TC0600001930.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600015099.mm.1 | NM_028462.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015099.mm.1 | NM_028462.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015099.mm.1 | TC0600001930.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429991_PM_at | - | - |
| Mouse Expression Set 430 | 1429991_at | - | - |
| Murine Genome U74v2 Set | 167382_PM_at | - | - |
| Murine Genome U74v2 Set | 167382_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429991_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429991_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 579295 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2733155 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2733155 | - | - |

Fezf1
Genomic Map