Assay Details
| Gene Symbol: Prl7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011168.4 | NP_035298.1 | 3 - 4 | 288 | 94 | |
| GenBank mRNA | AF011382.1 | - | 3 - 4 | 277 | 94 | |
| AF020524.1 | - | 3 - 4 | 284 | 94 | ||
| AK005455.1 | - | 3 - 4 | 289 | 94 | ||
| AK028068.1 | - | 3 - 4 | 288 | 94 | ||
| AK131665.1 | - | 3 - 4 | 289 | 94 | ||
| BC099468.1 | - | 3 - 4 | 267 | 30789902 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prl7a2
Genomic Map