Assay Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138589.2 | NP_613055.2 | 1 - 2 | 290 | 86 | |
| GenBank mRNA | AK017851.1 | - | 1 - 2 | 67 | 86 | |
| AK033428.1 | - | 1 - 2 | 281 | 86 | ||
| AK077395.1 | - | 1 - 2 | 313 | 86 | ||
| BC011313.1 | - | 1 - 2 | 293 | 3499738 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017851.1 transcripts.
Set Membership: |
|

Ubfd1
Genomic Map