Assay Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138589.2 | NP_613055.2 | 2 - 3 | 613 | 123 | |
| GenBank mRNA | AK011862.1 | - | 2 - 3 | 369 | 123 | |
| AK017851.1 | - | 2 - 3 | 390 | 123 | ||
| AK077395.1 | - | 2 - 3 | 636 | 123 | ||
| AK135258.1 | - | 2 - 3 | 371 | 123 | ||
| AK140891.1 | - | 2 - 3 | 372 | 123 | ||
| BC011313.1 | - | 2 - 3 | 616 | 3499738 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubfd1
Genomic Map