Assay Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138589.2 | NP_613055.2 | 4 - 5 | 888 | 81 | |
| GenBank mRNA | AK011862.1 | - | 4 - 5 | 644 | 81 | |
| AK033428.1 | - | 3 - 4 | 1667 | 81 | ||
| AK077395.1 | - | 4 - 5 | 911 | 81 | ||
| AK140891.1 | - | 4 - 5 | 647 | 81 | ||
| BC011313.1 | - | 4 - 5 | 891 | 3499738 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubfd1
Genomic Map