| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_001130188.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_001130188.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_001130189.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_001130189.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_001130190.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_001130190.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_001130191.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_001130191.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_011360.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
NM_011360.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600014251.mm.1 |
TC0600001802.mm.1 |
MTA TC |