Assay Details
| Gene Symbol: Mios | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145374.2 | NP_663349.2 | 4 - 5 | 1676 | 94 | |
| GenBank mRNA | BC020002.1 | - | 4 - 5 | 1674 | 3584335 | 94 |
| BC025652.1 | - | 2 - 3 | 744 | 94 | ||
| BQ876300.1 | - | 2 - 3 | 231 | 94 | ||
| CD807728.1 | - | 2 - 3 | 162 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mios
Genomic Map