Assay Details
| Gene Symbol: Gnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081143.1 | NP_001074612.1 | 6 - 7 | 829 | 104 | |
| GenBank mRNA | AK040065.1 | - | 6 - 7 | 830 | 104 | |
| BC147449.1 | - | 6 - 7 | 850 | 9056091 | 104 | |
| BC147466.1 | - | 6 - 7 | 850 | 9056108 | 104 | |
| BC147839.1 | - | 6 - 7 | 778 | 9087556 | 104 | |
| BC147841.1 | - | 6 - 7 | 778 | 9087558 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnat3
Genomic Map