Assay Details
| Gene Symbol: Asb15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080847.3 | NP_543123.2 | 3 - 4 | 284 | 111 | |
| GenBank mRNA | AF403043.1 | - | 3 - 4 | 285 | 111 | |
| AK019556.1 | - | 3 - 4 | 285 | 111 | ||
| AK029759.1 | - | 3 - 4 | 216 | 111 | ||
| AK079418.1 | - | 3 - 4 | 390 | 111 | ||
| BC050794.1 | - | 3 - 4 | 233 | 6742480 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asb15
Genomic Map