Assay Details
| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143755.1 | NP_001137227.1 | 1 - 2 | 79 | 98 | |
| GenBank mRNA | AI614174.1 | - | 1 - 2 | 79 | 98 | |
| AK006931.1 | - | 1 - 2 | 84 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001143755.1 transcripts.
Set Membership: |
|

Lrrc27
Genomic Map