Assay Details
| Gene Symbol: Hist1h4i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011560.1 | - | 1 | 668 | 118 | |
| BC019757.1 | - | 1 | 660 | 118 | ||
| BC052219.1 | - | 1 | 640 | 118 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hist1h4i
Genomic Map