Assay Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008944.2 | NP_032970.2 | 1 - 2 | 78 | 72 | |
| GenBank mRNA | AF099733.1 | - | 1 - 2 | 45 | 72 | |
| AK012119.1 | - | 1 - 2 | 60 | 72 | ||
| AK012990.1 | - | 1 - 2 | 67 | 72 | ||
| AK035578.1 | - | 67 | 72 | |||
| AK136115.1 | - | 1 - 2 | 80 | 72 | ||
| AK151883.1 | - | 1 - 2 | 69 | 72 | ||
| AK152862.1 | - | 1 - 2 | 69 | 72 | ||
| AK154011.1 | - | 1 - 2 | 62 | 72 | ||
| AK167752.1 | - | 1 - 2 | 69 | 72 | ||
| BC026768.1 | - | 1 - 2 | 51 | 3156907 | 72 | |
| CJ052982.1 | - | 1 - 2 | 80 | 72 | ||
| X70303.1 | - | 1 - 2 | 122 | 72 | ||
More Information
Set Membership: |
|

Psma2
Genomic Map