Assay Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008944.2 | NP_032970.2 | 4 - 5 | 406 | 86 | |
| GenBank mRNA | AF099733.1 | - | 4 - 5 | 373 | 86 | |
| AK012119.1 | - | 4 - 5 | 388 | 86 | ||
| AK012990.1 | - | 4 - 5 | 395 | 86 | ||
| AK035578.1 | - | 395 | 86 | |||
| AK136115.1 | - | 4 - 5 | 408 | 86 | ||
| AK151883.1 | - | 4 - 5 | 397 | 86 | ||
| AK152862.1 | - | 4 - 5 | 397 | 86 | ||
| AK154011.1 | - | 4 - 5 | 390 | 86 | ||
| AK167752.1 | - | 4 - 5 | 397 | 86 | ||
| BC026768.1 | - | 4 - 5 | 379 | 3156907 | 86 | |
| CJ052982.1 | - | 4 - 5 | 408 | 86 | ||
| X70303.1 | - | 4 - 5 | 450 | 86 | ||
More Information
Set Membership: |
|

Psma2
Genomic Map