Assay Details
| Gene Symbol: Braf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139294.5 | NP_647455.3 | 4 - 5 | 626 | 63 | |
| GenBank mRNA | AJ276307.1 | - | 2 - 3 | 310 | 63 | |
| AK140431.1 | - | 4 - 5 | 627 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Braf
Genomic Map