Assay Details
| Gene Symbol: Arhgef25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166413.1 | NP_001159885.1 | 11 - 12 | 1108 | 67 | |
| NM_028027.3 | NP_082303.2 | 11 - 12 | 1170 | 67 | ||
| GenBank mRNA | AF487515.1 | - | 11 - 12 | 1136 | 67 | |
| AK010452.1 | - | 11 - 12 | 1136 | 67 | ||
| AK158684.1 | - | 11 - 12 | 1108 | 67 | ||
| BC023367.1 | - | 2 - 3 | 147 | 67 | ||
| BC041113.1 | - | 9 - 10 | 1484 | 67 | ||
| BC054833.1 | - | 3 - 4 | 392 | 6439728 | 67 | |
More Information
Set Membership: |
|

Arhgef25
Genomic Map