| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000023130.mm.1 |
NM_030714.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023130.mm.1 |
NM_030714.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023130.mm.1 |
TC1000003104.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023126.mm.1 |
NM_030714.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023126.mm.1 |
NM_030714.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023126.mm.1 |
TC1000003104.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023127.mm.1 |
NM_030714.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023127.mm.1 |
NM_030714.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023127.mm.1 |
TC1000003104.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023128.mm.1 |
NM_030714.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023128.mm.1 |
NM_030714.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023128.mm.1 |
TC1000003104.mm.1 |
MTA TC |