Assay Details
| Gene Symbol: Erich5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173421.2 | NP_775597.1 | 2 - 3 | 1290 | 145 | |
| GenBank mRNA | BC030476.1 | - | 2 - 3 | 1298 | 5368170 | 145 |
| BC099414.1 | - | 2 - 3 | 1041 | 30931867 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144781 | BC030476.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144781 | NM_173421.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766569 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274993 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182426 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173358 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127342 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109566 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144781 | BC030476.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144781 | NM_173421.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30128640 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08331529 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109566 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127342 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03008027 | - | - |
| Whole Mouse Genome G4122A | A_51_P330825 | BC030476.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P330825 | NM_173421.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129388 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109566 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08331529 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127342 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12151620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12151609 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12151613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30128750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12255459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12148579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12160565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12233445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12051128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12148566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12255460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12051121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12460966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11879550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30128679 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10423625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10423628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10423627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10423624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10423626 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10423623 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500001777.mm.1 | NM_173421.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001777.mm.1 | NM_173421.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001777.mm.1 | TC1500000239.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500001778.mm.1 | NM_173421.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001778.mm.1 | NM_173421.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500001778.mm.1 | TC1500000239.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 652378 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6380148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006380148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2597343 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2597343 | - | - |
| mouse-6 v1.1 (Illumina) | scl47999.3_178-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6380148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47999.3_178-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6380148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2819380 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2597343 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2819380 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2597343 | - | - |

Erich5
Genomic Map