Assay Details
| Gene Symbol: Trpv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007572.2 | NP_001007573.1 | 7 - 8 | 954 | 81 | |
| GenBank mRNA | AK085479.1 | - | 7 - 8 | 1000 | 81 | |
| BC110554.1 | - | 7 - 8 | 954 | 40047811 | 81 | |
| BC110555.1 | - | 7 - 8 | 954 | 40047816 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpv5
Genomic Map