Assay Details
| Gene Symbol: Trpv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007572.2 | NP_001007573.1 | 9 - 10 | 1261 | 87 | |
| GenBank mRNA | AK085479.1 | - | 9 - 10 | 1307 | 87 | |
| BC110554.1 | - | 9 - 10 | 1261 | 40047811 | 87 | |
| BC110555.1 | - | 9 - 10 | 1261 | 40047816 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpv5
Genomic Map