Assay Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC094915.1 | - | 135 | 6435959 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC094915.1 transcripts.
Set Membership: |
|

Pou6f2
Genomic Map