Assay Details
| Gene Symbol: 1110038B12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027943.1 | 3 - 4 | 201 | 92 | ||
| GenBank mRNA | AK004150.1 | - | 3 - 4 | 202 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1110038B12Rik
Genomic Map