Assay Details
| Gene Symbol: Lrrc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028977.1 | NP_083253.1 | 3 - 4 | 1247 | 80 | |
| GenBank mRNA | AK018071.1 | - | 3 - 4 | 1213 | 80 | |
| BC030317.1 | - | 3 - 4 | 1166 | 5372009 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301930 | AK018071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301930 | BC030317.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301930 | CD741871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301930 | NM_028977.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301930 | AK018071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301930 | BC030317.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301930 | CD741871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301930 | NM_028977.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301930 | AK018071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301930 | BC030317.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301930 | CD741871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301930 | NM_028977.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301930 | AK018071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301930 | BC030317.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301930 | CD741871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301930 | NM_028977.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301930 | AK018071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301930 | BC030317.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301930 | CD741871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301930 | NM_028977.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301930 | AK018071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301930 | BC030317.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301930 | CD741871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301930 | NM_028977.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23526497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04876049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04519095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04483739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04952953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04421574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04623805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04641365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04250293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04641369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04623799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04421573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04463749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04719104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04914545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04483740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04250310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23526661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04598306 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10519999 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520000 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520001 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10520002 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429679_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454076_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528310 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10519998 | - | - |
| Mouse Genome 430 2.0 Array | 1429679_at | - | - |
| Mouse Genome 430 2.0 Array | 1429679_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500001238.mm.1 | NM_028977.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001238.mm.1 | NM_028977.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500001238.mm.1 | TC0500000173.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429679_PM_at | - | - |
| Mouse Expression Set 430 | 1429679_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454076_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429679_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454076_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429679_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 806634 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106290138 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2511456 | - | - |

Lrrc17
Genomic Map