Assay Details
| Gene Symbol: Slc26a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289787.1 | NP_001276716.1 | 1 - 2 | 107 | 60 | |
| NM_001289788.1 | NP_001276717.1 | 1 - 2 | 107 | 60 | ||
| NM_030727.5 | NP_109652.3 | 2 - 3 | 182 | 60 | ||
| GenBank mRNA | AF529192.1 | - | 2 - 3 | 182 | 60 | |
| BC108986.1 | - | 1 - 2 | 99 | 40047724 | 60 | |
| BC108987.1 | - | 1 - 2 | 99 | 40047725 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF529192.1, BC108986.1, BC108987.1, NM_001289787.1, NM_001289788.1, NM_030727.5 transcripts.
Set Membership: |
|

Slc26a5
Genomic Map