Assay Details
| Gene Symbol: Pom121l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162928.1 | NP_001156400.1 | 1 | 3436 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156806 | NM_001162929.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156806 | AK133022.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156810 | NM_001162928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979407 | NM_001162928.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 442690 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931351 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108081 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979407 | NM_001162928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156810 | NM_001162928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156806 | NM_001162929.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156806 | AK133022.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07807211 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07807222 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07807211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10779594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10636379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10636360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10636407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10636371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10636418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10636412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10636416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10636376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28875626 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10403974 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436975_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403973 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300001239.mm.1 | NM_001162928.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001239.mm.1 | NM_001162928.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001239.mm.1 | TC1300000187.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001238.mm.1 | NM_001162928.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001238.mm.1 | NM_001162928.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001238.mm.1 | TC1300000187.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436975_PM_at | - | - |
| Mouse Expression Set 430 | 1436975_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 516187 | - | - |

Pom121l2
Genomic Map