Assay Details
| Gene Symbol: Map7d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081124.1 | NP_001074593.1 | 3 - 4 | 559 | 63 | |
| GenBank mRNA | AK013466.1 | - | 3 - 4 | 350 | 63 | |
| AK162227.1 | - | 1 - 2 | 93 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK162227.1 transcripts.
Set Membership: |
|

Map7d2
Genomic Map