Assay Details
| Gene Symbol: Lrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172814.3 | NP_766402.1 | 5 - 6 | 1651 | 66 | |
| GenBank mRNA | AK050512.1 | - | 5 - 6 | 1652 | 66 | |
| AK084675.1 | - | 1 - 2 | 451 | 66 | ||
| BC058345.1 | - | 4 - 5 | 1831 | 6822657 | 66 | |
More Information
Set Membership: |
|

Lrp12
Genomic Map