Assay Details
| Gene Symbol: 4933400A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003635.1 | 1 - 2 | 115 | 73 | ||
| GenBank mRNA | AK016585.1 | - | 1 - 2 | 116 | 73 | |
| BC107240.1 | - | 1 - 2 | 37 | 73 | ||
| BC107241.1 | - | 1 - 2 | 37 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15245 | Gm15245 | OTTMUSG00000019597 | -302.2kb | Anti-sense | Chr. X - 169447830 - 169475913 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA G530011O06 gene | G530011O06Rik | 195.5kb | Sense | Chr. Y - 90751140 - 90755050 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016585.1 transcripts.
Set Membership: |
|

4933400A11Rik
Genomic Map